1,1'-Bis(di-i-propylphosphino)ferrocene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%

Supplier: Ambeed
Warning

A299272-100MG A299272-250MG
77250-768EA 110 USD
77250-768 77250-770
1,1'-Bis(di-i-propylphosphino)ferrocene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%
1,1'-Bis(di-i-propylphosphino)ferrocene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Precaución: for Research Use Only

Formula: C₃₀H₄₈BF₄FeP₂Rh
MW: 716.22 g/mol
MDL Number: MFCD02684572
CAS Number: 255064-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR