4-(2-Phenyl-5,7-bis(trifluoromethyl)pyrazolo[1,5-a]pyrimidin-3-yl)phenol 98%

Supplier: Ambeed
Danger

Synonyms: PHTPP

A299071-100MG A299071-10MG A299071-1MG A299071-25MG A299071-50MG A299071-5MG
77262-954EA 447.02 USD
77262-954 77262-956 77262-958 77262-960 77262-962 77262-964
4-(2-Phenyl-5,7-bis(trifluoromethyl)pyrazolo[1,5-a]pyrimidin-3-yl)phenol 98%
4-(2-Phenyl-5,7-bis(trifluoromethyl)pyrazolo[1,5-a]pyrimidin-3-yl)phenol

Precaución: for Research Use Only

Formula: C₂₀H₁₁F₆N₃O
MW: 423.31 g/mol
MDL Number: MFCD09971112
CAS Number: 805239-56-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR