(6-Chloropyridin-3-yl)(cyclopropyl)methanamine 97%

Supplier: Ambeed
Danger

A298905-1G A298905-250MG
77239-216EA 235 USD
77239-216 77239-218
(6-Chloropyridin-3-yl)(cyclopropyl)methanamine 97%
(6-Chloropyridin-3-yl)(cyclopropyl)methanamine

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂
MDL Number: MFCD18660953
CAS Number: 1270394-92-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR