4-(Carboxymethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A298475-1G A298475-250MG A298475-5G
77289-912EA 11 USD
77289-912 77289-914 77289-916
4-(Carboxymethoxy)benzoic acid 97%
4-(Carboxymethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₅
MDL Number: MFCD00004307
CAS Number: 19360-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR