5-Bromo-4,6-dihydroxypyrimidine 97%

Supplier: Ambeed
Warning

A298473-100G A298473-10G A298473-25G A298473-5G
77289-904EA 143 USD
77289-904 77289-906 77289-908 77289-910
5-Bromo-4,6-dihydroxypyrimidine 97%
5-Bromo-4,6-dihydroxypyrimidine

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MDL Number: MFCD00047380
CAS Number: 15726-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR