N-Acetyl-di-O-acetylganciclovir 97%

Supplier: Ambeed
Warning

Synonyms: 2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)propane-1,3-diyl diacetate, Triacetylganciclovir, N-[9-[[2-(Acetyloxy)-1-[(acetyloxy)methyl]ethoxy]methyl]-6,9-dihydro-6-oxo-1H-purin-2-yl]acetamide

A298472-100G A298472-10G A298472-25G A298472-5G
77239-204EA 192.61 USD
77239-204 77239-206 77239-208 77239-210
N-Acetyl-di-O-acetylganciclovir 97%
N-Acetyl-di-O-acetylganciclovir

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₅O₇
MW: 381.35 g/mol
MDL Number: MFCD08457700
CAS Number: 86357-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR