3,5-Dinitrobenzonitrile 98%

Supplier: Ambeed
Warning

A298458-1G A298458-25G A298458-5G
77277-280EA 13 USD
77277-280 77277-282 77277-284
3,5-Dinitrobenzonitrile 98%
3,5-Dinitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃N₃O₄
MDL Number: MFCD00007228
CAS Number: 4110-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR