2-Fluoro-m-cresol 98%

Supplier: Ambeed
Danger

A298446-100G A298446-1G A298446-5G A298446-25G
77262-944EA 455 USD
77262-944 77262-946 77262-948 77748-578
2-Fluoro-m-cresol 98%
2-Fluoro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₇FO
MDL Number: MFCD09835186
CAS Number: 77772-72-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR