Sodium-1-naphthol-8-sulfonate 96% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Sodium 8-Hydroxy-1-naphthalenesulphonate, Sodium 8-hydroxynaphthalene-1-sulphonate, 8-Hydroxy-1-naphthalenesulphonic acid Sodium Salt, 1-Naphthol-8-sulphonic acid Sodium Salt

A298266-1G A298266-5G
77239-200EA 5.15 USD
77239-200 77239-202
Sodium-1-naphthol-8-sulfonate 96% (by HPLC)
Sodium-1-naphthol-8-sulfonate

Precaución: for Research Use Only

Formula: C₁₀H₈O₄S
MW: 246.21 g/mol
MDL Number: MFCD00021524
CAS Number: 20215-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR