2-Chlor-4-fluorothiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chlor-4-fluorobenzene-1-thiol, 2-Chloro-4-fluorobenzenethiol, 1-Chloro-3-fluoro-6-mercaptobenzene, 2-Chlor-4-fluorobenzenethiol

A298050-5G A298050-25G
77262-932EA 62.83 USD
77262-932 77550-806
2-Chlor-4-fluorothiophenol 98%
2-Chlor-4-fluorothiophenol

Precaución: for Research Use Only

Formula: C₆H₄ClFS
MW: 162.61 g/mol
MDL Number: MFCD00052936
CAS Number: 175277-99-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR