3-Iodobenzo[b]thiophene 95%

Supplier: Ambeed
Warning

A297870-1G A297870-5G
77277-276EA 41.2 USD
77277-276 77277-278
3-Iodobenzo[b]thiophene 95%
3-Iodobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅IS
MW: 260.09 g/mol
MDL Number: MFCD00176396
CAS Number: 36748-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR