3,4-Xylylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethylphenylacetic acid

A297551-1G A297551-25G A297551-5G
77277-268EA 21.63 USD
77277-268 77277-270 77277-272
3,4-Xylylacetic acid 97%
3,4-Xylylacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD02664684
CAS Number: 17283-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR