1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 96%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-Propylpyrazole-4-boronic acid Pinacol ester, 2-(1-Propyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Propyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
A297285-100MG
A297285-250MG
A297285-5G
A297285-1G
77239-184EA
5
USD
77239-184
77239-186
77239-188
77746-559
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 96%
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₁₂H₂₁BN₂O₂ |
MDL Number:
MFCD05663871 CAS Number: 827614-69-7 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...