4'-Methylacetanilide 96%

Supplier: Ambeed
Warning

Synonyms: p-Acetotoluidid, p-Acetotoluidide, N-(4-Methylphenyl)acetamide

A297072-100G A297072-10G A297072-25G A297072-5G
77289-896EA 55.62 USD
77289-896 77289-898 77289-900 77289-902
4'-Methylacetanilide 96%
4'-Methylacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD00008677
CAS Number: 103-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR