1-Methyl-1H-indole-6-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A297018-1G
A297018-100MG
77239-182EA
79.31
USD
77239-182
77746-555
1-Methyl-1H-indole-6-carboxylic acid 95%
1-Methyl-1H-indole-6-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₂ |
MDL Number:
MFCD08690250 CAS Number: 202745-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...