3-Chloro-1-phenylpropan-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-1-phenyl-1-propanol, 3-Chloro-1-phenyl-propanol, alpha-(2-Chloroethyl)benzyl alcohol

A296595-100G A296595-10G A296595-1G A296595-25G A296595-5G
77277-254EA 164 USD
77277-254 77277-256 77277-258 77277-260 77277-262
3-Chloro-1-phenylpropan-1-ol 97%
3-Chloro-1-phenylpropan-1-ol

Precaución: for Research Use Only

Formula: C9H11ClO
MW: 170.6400 g/mol
MDL Number: MFCD09878800
CAS Number: 18776-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR