1-Methoxy-2-methyl-2-propanol 95%

Supplier: Ambeed
Danger

A296401-1G A296401-250MG
77239-176EA 9 USD
77239-176 77239-178
1-Methoxy-2-methyl-2-propanol 95%
1-Methoxy-2-methyl-2-propanol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MDL Number: MFCD03701584
CAS Number: 3587-64-2
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR