2,6,10-Trimethyldodecane 98%

Supplier: Ambeed
Danger

A296369-100MG A296369-1G A296369-250MG
77250-752EA 204.01 USD
77250-752 77250-754 77250-756
2,6,10-Trimethyldodecane 98%
2,6,10-Trimethyldodecane

Precaución: for Research Use Only

Formula: C₁₅H₃₂
MW: 212.415 g/mol
MDL Number: MFCD00027077
CAS Number: 3891-98-3
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR