4-Fluoro-3-nitrobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A296253-100G A296253-10G A296253-1G A296253-250MG A296253-25G A296253-5G
77289-884EA 247 USD
77289-884 77289-886 77289-888 77289-890 77289-892 77289-894
4-Fluoro-3-nitrobenzenesulfonamide 98%
4-Fluoro-3-nitrobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₄S
MW: 220.18 g/mol
MDL Number: MFCD08703185
CAS Number: 406233-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR