Sodium 1-benzyl-1H-indazol-3-olate 97%

Supplier: Ambeed
Warning

A295943-10G A295943-1G A295943-250MG
77239-146EA 13.39 USD
77239-146 77239-148 77239-150
Sodium 1-benzyl-1H-indazol-3-olate 97%
Sodium 1-benzyl-1H-indazol-3-olate

Precaución: for Research Use Only

Formula: C₁₄H₁₁N₂NaO
MW: 246.24 g/mol
MDL Number: MFCD08458863
CAS Number: 13185-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR