tert-Butyl 3-benzyl-3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%

Supplier: Ambeed
Warning

A294278-100MG A294278-1G
77262-900EA 18.54 USD
77262-900 77262-902
tert-Butyl 3-benzyl-3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%
tert-Butyl 3-benzyl-3,6-diazabicyclo[3.1.1]heptane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₂
MW: 288.39 g/mol
MDL Number: MFCD30471713
CAS Number: 869494-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR