2-Methylveratraldehyde 97%

Supplier: Ambeed
Warning

A294237-100MG A294237-1G A294237-250MG
77239-110EA 25 USD
77239-110 77239-112 77239-114
2-Methylveratraldehyde 97%
2-Methylveratraldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD03428551
CAS Number: 51234-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR