4-Ethoxyphenyl trans-4-butylcyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Trans-4-ethoxyphenyl 4-butylcyclohexanecarboxylate, trans-4-Butylcyclohexanecarboxylic Acid 4-Ethoxyphenyl Ester

A294177-10G A294177-25G A294177-5G
77289-844EA 49.44 USD
77289-844 77289-846 77289-848
4-Ethoxyphenyl trans-4-butylcyclohexanecarboxylate 97%
4-Ethoxyphenyl trans-4-butylcyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₈O₃
MW: 304.43 g/mol
MDL Number: MFCD01941073
CAS Number: 67589-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR