2-(Difluoromethoxy)phenylacetonitrile 96%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(2-(Difluoromethoxy)phenyl)acetonitrile
A294089-1G
A294089-25G
77262-896EA
12.36
USD
77262-896
77262-898
2-(Difluoromethoxy)phenylacetonitrile 96%
2-(Difluoromethoxy)phenylacetonitrile
Precaución: for Research Use Only
Formula:
C₉H₇F₂NO MW: 183.1548 g/mol |
MDL Number:
MFCD00236240 CAS Number: 41429-22-5 UN: 3276 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...