Methyl 4-amino-2-isopropoxybenzoate 97%

Supplier: Ambeed
Warning

A293886-1G A293886-250MG
77250-724EA 202.91 USD
77250-724 77250-726
Methyl 4-amino-2-isopropoxybenzoate 97%
Methyl 4-amino-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.24 g/mol
MDL Number: MFCD24624427
CAS Number: 909563-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR