3-Mercapto-1-propanol 98%

Supplier: Ambeed
Danger

A293814-25G A293814-5G
77277-238EA 52 USD
77277-238 77277-240
3-Mercapto-1-propanol 98%
3-Mercapto-1-propanol

Precaución: for Research Use Only

Formula: C₃H₈OS
MDL Number: MFCD00192262
CAS Number: 19721-22-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR