6-Hydroxy-3,4-dihydroquinolin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydro-6-hydroxy-2(1H)-quinolinone, 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, Cilostazol Related Compound A, 6-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline

A293810-10G A293810-25G A293810-500G A293810-5G
77277-230EA 6.18 USD
77277-230 77277-232 77277-234 77277-236
6-Hydroxy-3,4-dihydroquinolin-2(1H)-one 98%
6-Hydroxy-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.1700 g/mol
MDL Number: MFCD02179410
CAS Number: 54197-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR