1-Bromododecane 98%

Supplier: Ambeed
Warning

A293791-100G A293791-25G A293791-500G
77239-102EA 7 USD
77239-102 77239-104 77239-106
1-Bromododecane 98%
1-Bromododecane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₁₀CH₃
MDL Number: MFCD00000225
CAS Number: 143-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR