2-Chloro-6-iodobenzonitrile 97%

Supplier: Ambeed
Warning

A293740-100MG A293740-1G A293740-250MG
77262-890EA 12.36 USD
77262-890 77262-892 77262-894
2-Chloro-6-iodobenzonitrile 97%
2-Chloro-6-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClIN
MW: 263.463 g/mol
MDL Number: MFCD01763605
CAS Number: 89642-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR