(1-Methylallyl)palladium chloride dimer 98%

Supplier: Ambeed
Warning

A293592-100MG A293592-1G A293592-250MG
77250-718EA 23 USD
77250-718 77250-720 77250-722
(1-Methylallyl)palladium chloride dimer 98%
(1-Methylallyl)palladium chloride dimer

Precaución: for Research Use Only

Formula: C₈H₁₄Cl₂Pd₂
MDL Number: MFCD01074457
CAS Number: 12081-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR