4-(Cyanomethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Cyanomethyl)benzeneboronic acid

A293566-1G A293566-250MG A293566-5G
77239-094EA 16 USD
77239-094 77239-096 77239-098
4-(Cyanomethyl)phenylboronic acid 98%
4-(Cyanomethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
MW: 160.97 g/mol
MDL Number: MFCD01632200
CAS Number: 91983-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR