(R)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate 97%

Supplier: Ambeed
Warning

A293307-10G A293307-1G A293307-25G A293307-5G
77239-086EA 27.81 USD
77239-086 77239-088 77239-090 77239-092
(R)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate 97%
(R)-2-Amino-4-oxo-4-(tritylamino)butanoic acid hydrate

Precaución: for Research Use Only

Formula: C₂₃H₂₄N₂O₄
MW: 392.45 g/mol
MDL Number: MFCD00153423
CAS Number: 1998701-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR