6-Bromochromone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-4H-chromen-4-one

A293200-1G A293200-25G
77262-886EA 15.45 USD
77262-886 77262-888
6-Bromochromone 98%
6-Bromochromone

Precaución: for Research Use Only

Formula: C₉H₅BrO₂
MW: 225.0388 g/mol
MDL Number: MFCD00239369
CAS Number: 51483-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR