2-Ethyl-5(or 6)-methylpyrazine (mixture of isomers) 95%
Supplier: Ambeed
|
Warning
|
A293112-100G
A293112-10G
A293112-1G
A293112-25G
A293112-5G
77262-876EA
189
USD
77262-876
77262-878
77262-880
77262-882
77262-884
2-Ethyl-5(or 6)-methylpyrazine (mixture of isomers) 95%
2-Ethyl-5(or 6)-methylpyrazine (mixture of isomers)
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂ |
MDL Number:
MFCD09039261 CAS Number: 36731-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...