2-Ethynyl-5-methoxypyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethynyl-5-methoxy-1,4-diazine, (5-Methoxypyrazin-2-yl)acetylene

A293045-100MG A293045-250MG
77250-710EA 55 USD
77250-710 77250-712
2-Ethynyl-5-methoxypyrazine 97%
2-Ethynyl-5-methoxypyrazine

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.135 g/mol
MDL Number: MFCD22199320
CAS Number: 1374115-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR