3-(2-Methylquinolin-1-ium-1-yl)propane-1-sulfonate 97%

Supplier: Ambeed
Warning

A293044-100G A293044-10G A293044-1G A293044-25G A293044-5G
77262-866EA 175.1 USD
77262-866 77262-868 77262-870 77262-872 77262-874
3-(2-Methylquinolin-1-ium-1-yl)propane-1-sulfonate 97%
3-(2-Methylquinolin-1-ium-1-yl)propane-1-sulfonate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃S
MW: 265.33 g/mol
MDL Number: MFCD00182042
CAS Number: 51583-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR