3,5-Dibromo-4-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A292879-250MG A292879-25G A292879-5G
77239-080EA 13 USD
77239-080 77239-082 77239-084
3,5-Dibromo-4-methylbenzoic acid 97%
3,5-Dibromo-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MDL Number: MFCD00017544
CAS Number: 67973-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR