2-Acetamido-5-pyridineboronic acid pinacol ester 95+%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamidopyridine-5-boronic acid, pinacol ester, N-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide

A292759-1G A292759-250MG A292759-5G
77250-702EA 91 USD
77250-702 77250-704 77250-706
2-Acetamido-5-pyridineboronic acid pinacol ester 95+%
2-Acetamido-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BN₂O₃
MW: 262.12 g/mol
MDL Number: MFCD08690229
CAS Number: 904326-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR