4-(Methylamino)benzonitrile 97%

Supplier: Ambeed
Warning

A292749-10G A292749-1G A292749-250MG A292749-25G A292749-5G
77289-812EA 29 USD
77289-812 77289-814 77289-816 77289-818 77289-820
4-(Methylamino)benzonitrile 97%
4-(Methylamino)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD00016381
CAS Number: 4714-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR