(S)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, (S)-(-)-DM-SEGPHOS®, (S)-(-)-4,4'-Bis[di(3,5-xylyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene)

A292621-100MG A292621-1G A292621-250MG A292621-5G
77239-074EA 32 USD
77239-074 77239-076 77239-078 77746-544
(S)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole 98%
(S)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₄₆H₄₄O₄P₂
MW: 722.790 g/mol
MDL Number: MFCD09753008
CAS Number: 210169-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR