Methyl-4-amino-3-hydroxybenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-amino-3-hydroxybenzoate
A292513-100G
A292513-10G
A292513-1G
A292513-25G
A292513-5G
77250-692EA
356.38
USD
77250-692
77250-694
77250-696
77250-698
77250-700
Methyl-4-amino-3-hydroxybenzoate 97%
Methyl 4-amino-3-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₉NO₃ MW: 167.17 g/mol |
MDL Number:
MFCD00017093 CAS Number: 63435-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...