Se-Phenyl-L-selenocysteine 97%

Supplier: Ambeed
Danger

A292053-100MG A292053-1G A292053-250MG
77239-068EA 130 USD
77239-068 77239-070 77239-072
Se-Phenyl-L-selenocysteine 97%
Se-Phenyl-L-selenocysteine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂Se
MDL Number: MFCD03791083
CAS Number: 71128-82-0
UN: 3283
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR