2,5-Bis(prop-2-yn-1-yloxy)terephthalaldehyde 97%

Supplier: Ambeed
Warning

A291938-100MG A291938-250MG A291938-1G
77262-834EA 50.47 USD
77262-834 77262-836 77550-772
2,5-Bis(prop-2-yn-1-yloxy)terephthalaldehyde 97%
2,5-Bis(prop-2-yn-1-yloxy)terephthalaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
MDL Number: MFCD31700807
CAS Number: 1538579-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR