Ethyl-7-hydroxycoumarin-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate, YZ9

A291718-1MG A291718-250MG A291718-1G
77277-174EA 5.15 USD
77277-174 77277-176 77748-566
Ethyl-7-hydroxycoumarin-3-carboxylate 97%
Ethyl-7-hydroxycoumarin-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₅
MW: 234.21 g/mol
MDL Number: MFCD00017641
CAS Number: 6093-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR