MSC-2530818 98+%

Supplier: Ambeed
Warning

Synonyms: MSC2530818, (S)-(2-(4-Chlorophenyl)pyrrolidin-1-yl)(3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)methanone

A291701-10MG A291701-1MG A291701-5MG A291701-50MG
77262-828EA 35.02 USD
77262-828 77262-830 77262-832 77748-565
MSC-2530818 98+%
MSC-2530818

Precaución: for Research Use Only

Formula: C₁₈H₁₇ClN₄O
MW: 340.81 g/mol
MDL Number: MFCD31382190
CAS Number: 1883423-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR