(1S)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1S)-1-(diphenylphosphino)ethyl]ferrocene ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A291442-100MG
A291442-1G
A291442-250MG
A291442-5G
77250-676EA
21
USD
77250-676
77250-678
77250-680
77250-682
(1S)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1S)-1-(diphenylphosphino)ethyl]ferrocene ≥98%, ee 99%
(1S)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1S)-1-(diphenylphosphino)ethyl]ferrocene
Precaución: for Research Use Only
Formula:
C₃₂H₄₀FeP₂ MW: 542.45 g/mol |
MDL Number:
MFCD08561171 CAS Number: 223121-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...