2-Isopropoxyphenol 97%

Supplier: Ambeed
Warning

A290965-1G A290965-25G A290965-5G
77262-818EA 6.18 USD
77262-818 77262-820 77262-822
2-Isopropoxyphenol 97%
2-Isopropoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00002184
CAS Number: 4812-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR