(R)-(+)-1,1'-Bi-2-naphthol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-[1,1'-Binaphthalene]-2,2'-diol, (R)-BINOL, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol
A290939-10G
A290939-25G
A290939-5G
77289-790EA
9.27
USD
77289-790
77289-792
77289-794
(R)-(+)-1,1'-Bi-2-naphthol 98%
(R)-(+)-1,1'-Bi-2-naphthol
Precaución: for Research Use Only
Formula:
C₂₀H₁₄O₂ MW: 286.33 g/mol |
MDL Number:
MFCD00004068 CAS Number: 18531-94-7 UN: 2811 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...