3',5'-Dibromo-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

A290775-1G A290775-250MG A290775-5G
77239-062EA 8 USD
77239-062 77239-064 77239-066
3',5'-Dibromo-2'-hydroxyacetophenone 98%
3',5'-Dibromo-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MDL Number: MFCD00191992
CAS Number: 22362-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR