2-Amino-3-methylpyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methylpyrazin-2-amine

A290517-10G A290517-1G A290517-250MG A290517-25G A290517-5G
77250-660EA 203.94 USD
77250-660 77250-662 77250-664 77250-666 77250-668
2-Amino-3-methylpyrazine 97%
2-Amino-3-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.132 g/mol
MDL Number: MFCD08705770
CAS Number: 19838-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR